Assay Details
Target Gene Details
Entrez Gene ID: | 100144595 |
Gene Name: | uncharacterized LOC100144595 |
Gene Aliases: |
- |
Location: |
Chr.2:154435853-154457438 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100144595 | NM_001256368.1 | 2 | 144 | NP_001243297.1 |
| AK090913.1 | 2 | 145 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv583421 | Chr.2:154401008 - 154524026 on Build GRCh38 | Loss |
|
| nsv583422 | Chr.2:154406489 - 154475974 on Build GRCh38 | Loss |
|
| esv3592993 | Chr.2:154447556 - 154553185 on Build GRCh38 | Gain |
|

Genomic Map