Assay Details
Target Gene Details
Entrez Gene ID: | 151393 |
Gene Name: | regulator of microtubule dynamics 2 |
Gene Aliases: |
BLOCK18, FAM82A, FAM82A1, PRO34163, PYST9371, RMD-2, RMD2, RMD4 |
Location: |
Chr.2:37920789-38068032 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RMDN2 | NM_001170791.2 | NP_001164262.1 | ||
| NM_001170792.2 | NP_001164263.1 | |||
| NM_001322211.1 | NP_001309140.1 | |||
| NM_001322212.1 | NP_001309141.1 | |||
| XM_011532614.2 | XP_011530916.1 | |||
| XM_011532615.2 | XP_011530917.2 | |||
| XM_011532616.2 | XP_011530918.1 | |||
| XM_011532617.2 | XP_011530919.1 | |||
| XM_011532618.2 | XP_011530920.1 | |||
| XM_011532619.2 | XP_011530921.1 | |||
| XM_017003474.1 | XP_016858963.1 | |||
| XM_017003475.1 | XP_016858964.1 | |||
| XM_017003476.1 | XP_016858965.1 | |||
| XM_017003477.1 | XP_016858966.1 | |||
| XM_017003478.1 | XP_016858967.1 | |||
| XM_017003479.1 | XP_016858968.1 | |||
| AK095462.1 | BAC04551.1 | |||
| AY358269.1 | AAQ88636.1 | |||
| BC024243.2 | AAH24243.3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv6787n54 | Chr.2:37855471 - 37965002 on Build GRCh38 | Loss |
|
| nsv963223 | Chr.2:37930010 - 37931922 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map