Assay Details
Target Gene Details
Entrez Gene ID: | 150684 |
Gene Name: | copper metabolism domain containing 1 |
Gene Aliases: |
C2orf5, MURR1 |
Location: |
Chr.2:61888723-62136070 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COMMD1 | NM_001321781.1 | NP_001308710.1 | ||
| NM_001321782.1 | NP_001308711.1 | |||
| DT217730.1 | ||||
| HY039942.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3591027 | Chr.2:61889450 - 61893138 on Build GRCh38 | Loss |
|
| nsv582123 | Chr.2:61719360 - 62324337 on Build GRCh38 | Gain |
|
| nsv458074 | Chr.2:61692252 - 62020316 on Build GRCh38 | Gain |
|
| nsv470467 | Chr.2:61786803 - 62020316 on Build GRCh38 | Gain |
|
| nsv582118 | Chr.2:61431550 - 62075525 on Build GRCh38 | Gain |
|
| esv34000 | Chr.2:61672382 - 62157212 on Build GRCh38 | Loss |
|
| nsv1009446 | Chr.2:61872503 - 61972765 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76353526] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map