Assay Details
Target Gene Details
Entrez Gene ID: | 64342 |
Gene Name: | HCLS1 binding protein 3 |
Gene Aliases: |
ETM2, HS1-BP3 |
Location: |
Chr.2:20553861-20651661 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HS1BP3 | XM_017004696.1 | XP_016860185.1 | ||
| XM_017004697.1 | XP_016860186.1 | |||
| XM_017004698.1 | XP_016860187.1 | |||
| XM_017004699.1 | XP_016860188.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953754 | Chr.2:20558041 - 20574140 on Build GRCh38 | Deletion |
|
| dgv56n68 | Chr.2:20430331 - 20664318 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map