Assay Details
Target Gene Details
Entrez Gene ID: | 8809 |
Gene Name: | interleukin 18 receptor 1 |
Gene Aliases: |
CD218a, CDw218a, IL-1Rrp, IL18RA, IL1RRP |
Location: |
Chr.2:102355817-102398777 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL18R1 | NM_001282399.1 | NP_001269328.1 | ||
| NM_003855.3 | NP_003846.1 | |||
| XM_005264039.3 | XP_005264096.1 | |||
| XM_005264040.3 | XP_005264097.1 | |||
| XM_011512099.1 | XP_011510401.1 | |||
| XM_017005181.1 | XP_016860670.1 | |||
| XM_017005182.1 | XP_016860671.1 | |||
| XM_017005183.1 | XP_016860672.1 | |||
| XM_017005184.1 | XP_016860673.1 | |||
| AY192162.1 | AAO41718.1 | |||
| BC069575.1 | AAH69575.1 | |||
| BC093975.1 | AAH93975.1 | |||
| BC093977.1 | AAH93977.1 | |||
| BC143403.1 | ||||
| DR006178.1 | ||||
| U43672.1 | AAC50390.1 |
More Information
Set Membership: |
|

Genomic Map