Assay Details
Target Gene Details
Entrez Gene ID: | 1796 |
Gene Name: | docking protein 1 |
Gene Aliases: |
P62DOK, pp62 |
Location: |
Chr.2:74549020-74557551 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DOK1 | NM_001197260.1 | NP_001184189.1 | ||
| NM_001318866.1 | NP_001305795.1 | |||
| NM_001318867.1 | NP_001305796.1 | |||
| NM_001318868.1 | NP_001305797.1 | |||
| NM_001318869.1 | NP_001305798.1 | |||
| NM_001381.4 | NP_001372.1 | |||
| XM_017003528.1 | 3 | 1097 | XP_016859017.1 | |
| AF035299.1 | AAB88182.1 | |||
| AF180527.1 | AAF19167.1 | |||
| AK055944.1 | ||||
| AK127127.1 | 3 | 872 | ||
| AK127656.1 | 3 | 872 | ||
| AK222883.1 | BAD96603.1 | |||
| AK296154.1 | ||||
| AK300097.1 | ||||
| AK314678.1 | ||||
| BC111055.1 | AAI11056.1 | |||
| BC114440.1 | ||||
| BC114497.1 | ||||
| U70987.1 | AAC51127.1 |
More Information
Set Membership: |
|

Genomic Map