Assay Details
Target Gene Details
Entrez Gene ID: | 11321 |
Gene Name: | GPN-loop GTPase 1 |
Gene Aliases: |
ATPBD1A, MBDIN, NTPBP, RPAP4, XAB1 |
Location: |
Chr.2:27628648-27650846 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPN1 | NM_001145047.1 | NP_001138519.1 | ||
| NM_001145048.1 | NP_001138520.1 | |||
| NM_001145049.1 | NP_001138521.1 | |||
| NM_007266.3 | NP_009197.2 | |||
| NR_026735.1 | ||||
| AB044661.1 | BAB17612.1 | |||
| AJ010842.1 | CAA09376.1 | |||
| AK298827.1 | ||||
| AK298866.1 | ||||
| AK302131.1 | ||||
| AK310340.1 | ||||
| AK312992.1 | ||||
| AK316404.1 | ||||
| BC007451.2 | AAH07451.1 |
Target Gene Details
Entrez Gene ID: | 9913 |
Gene Name: | SPT7-like STAGA complex gamma subunit |
Gene Aliases: |
SPT7L, STAF65, STAF65(gamma), STAF65G, SUPT7H |
Location: |
Chr.2:27642568-27663840 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SUPT7L |
More Information
Set Membership: |
|

Genomic Map