Assay Details
Target Gene Details
Entrez Gene ID: | 1796 |
Gene Name: | docking protein 1 |
Gene Aliases: |
P62DOK, pp62 |
Location: |
Chr.2:74549020-74557551 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DOK1 | NM_001197260.1 | NP_001184189.1 | ||
| AF035299.1 | AAB88182.1 | |||
| DA382785.1 |
Target Gene Details
Entrez Gene ID: | 84695 |
Gene Name: | lysyl oxidase like 3 |
Gene Aliases: |
LOXL |
Location: |
Chr.2:74532258-74555709 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOXL3 | NM_001289164.1 | NP_001276093.1 | ||
| NM_032603.3 | NP_115992.1 | |||
| XM_011533134.2 | XP_011531436.1 | |||
| XM_017005112.1 | XP_016860601.1 | |||
| AF282619.1 | AAK51671.1 | |||
| AF284815.1 | AAK91134.1 | |||
| AF311313.1 | AAK63205.1 | |||
| AK309474.1 | ||||
| AK309478.1 | ||||
| BC071865.1 | AAH71865.1 | |||
| DA554977.1 |
More Information
Set Membership: |
|

Genomic Map