Assay Details
Target Gene Details
Entrez Gene ID: | 25904 |
Gene Name: | CCR4-NOT transcription complex subunit 10 |
Gene Aliases: |
- |
Location: |
Chr.3:32685145-32773875 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNOT10 | NM_001256741.1 | NP_001243670.1 | ||
| NM_015442.2 | NP_056257.1 | |||
| NR_046352.1 | ||||
| XM_017006109.1 | XP_016861598.1 | |||
| XM_017006110.1 | XP_016861599.1 | |||
| XM_017006111.1 | XP_016861600.1 | |||
| XM_017006112.1 | XP_016861601.1 | |||
| AK022576.1 | BAB14108.1 | |||
| AK022952.1 | BAB14327.1 | |||
| AK023227.1 | BAB14478.1 | |||
| AK297085.1 | ||||
| AK298693.1 | ||||
| AK309670.1 | ||||
| BC002928.2 | AAH02928.1 | |||
| BC002931.2 | AAH02931.1 | |||
| DB107280.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003285 | Chr.3:31386933 - 33172886 on Build GRCh38 | Gain |
|
| nsv998302 | Chr.3:32595752 - 33112754 on Build GRCh38 | Gain |
|
| nsv1004530 | Chr.3:32623361 - 32719438 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map