Assay Details
Target Gene Details
Entrez Gene ID: | 102724604 |
Gene Name: | uncharacterized LOC102724604 |
Gene Aliases: |
- |
Location: |
Chr.3:181421163-181442486 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102724604 | NR_125407.1 | |||
| DA352137.1 |
Target Gene Details
Entrez Gene ID: | 347689 |
Gene Name: | SOX2 overlapping transcript |
Gene Aliases: |
NCRNA00043, SOX2OT |
Location: |
Chr.3:181056680-181742228 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOX2-OT | NR_075091.1 | |||
| NR_075092.1 | ||||
| NR_075093.1 | ||||
| DA281835.1 | ||||
| DA310380.1 | ||||
| JQ408703.1 | ||||
| JX110620.1 | ||||
| JX504683.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829810 | Chr.3:181401148 - 181573603 on Build GRCh38 | Gain+Loss |
|

Genomic Map