Assay Details
Target Gene Details
Entrez Gene ID: | 105374205 |
Gene Name: | uncharacterized LOC105374205 |
Gene Aliases: |
- |
Location: |
Chr.3:169447867-169477052 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105374205 | NR_134932.1 |
Target Gene Details
Entrez Gene ID: | 2122 |
Gene Name: | MDS1 and EVI1 complex locus |
Gene Aliases: |
AML1-EVI-1, EVI1, MDS1, MDS1-EVI1, PRDM3, RUSAT2 |
Location: |
Chr.3:169083499-169663781 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MECOM | NM_001205194.1 | NP_001192123.1 | ||
| NM_004991.3 | NP_004982.2 | |||
| XM_005247213.3 | XP_005247270.1 | |||
| XM_005247214.3 | XP_005247271.1 | |||
| XM_005247215.3 | XP_005247272.1 | |||
| XM_005247224.3 | XP_005247281.1 | |||
| XM_005247225.3 | XP_005247282.1 | |||
| XM_005247226.3 | XP_005247283.1 | |||
| XM_017005877.1 | XP_016861366.1 | |||
| AK304098.1 | ||||
| AK308404.1 | ||||
| AL700380.1 | ||||
| BC069498.1 | AAH69498.1 | |||
| CR541866.1 | CAG46664.1 | |||
| CR541886.1 | CAG46684.1 | |||
| DC347146.1 | ||||
| DC348381.1 | ||||
| GQ352634.1 | ||||
| U43292.1 | AAB05839.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829792 | Chr.3:169425780 - 169570628 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs80131709] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map