Assay Details
Target Gene Details
Entrez Gene ID: | 100874092 |
Gene Name: | UBE2E1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:23804024-23806905 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE2E1-AS1 | NR_046652.1 | 2 | 181 | |
| ES608176.1 | 2 | 181 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834637 | Chr.3:23662437 - 23844653 on Build GRCh38 | Loss |
|
| nsv1005186 | Chr.3:21989661 - 24089392 on Build GRCh38 | Gain |
|
| esv3575556 | Chr.3:23642287 - 23815715 on Build GRCh38 | Gain |
|

Genomic Map