Assay Details
Target Gene Details
Entrez Gene ID: | 100507524 |
Gene Name: | ARHGEF26 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:154024401-154121210 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGEF26-AS1 | NR_037901.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs142390784] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map