Assay Details
Target Gene Details
Entrez Gene ID: | 55802 |
Gene Name: | decapping mRNA 1A |
Gene Aliases: |
HSA275986, Nbla00360, SMAD4IP1, SMIF |
Location: |
Chr.3:53283423-53347627 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCP1A | NM_001290204.1 | 9 | 5304 | NP_001277133.1 |
| NM_001290205.1 | 11 | 5473 | NP_001277134.1 | |
| NM_001290206.1 | 9 | 5290 | NP_001277135.1 | |
| NM_001290207.1 | 8 | 5176 | NP_001277136.1 | |
| NM_018403.6 | 10 | 5418 | NP_060873.4 | |
| AK025344.1 | 1 | 1214 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834699 | Chr.3:53117860 - 53291778 on Build GRCh38 | Loss |
|
| nsv517233 | Chr.3:53141541 - 53286984 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map