Assay Details
Target Gene Details
Entrez Gene ID: | 5449 |
Gene Name: | POU class 1 homeobox 1 |
Gene Aliases: |
CPHD1, GHF-1, PIT1, POU1F1a, Pit-1 |
Location: |
Chr.3:87259633-87276587 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POU1F1 | NM_000306.3 | NP_000297.1 | ||
| NM_001122757.2 | NP_001116229.1 | |||
| D10216.1 | BAA01068.1 | |||
| L18781.1 | AAA60093.1 | |||
| X62429.1 | CAA44295.1 | |||
| X72215.1 | CAA51017.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527279 | Chr.3:86796784 - 87814130 on Build GRCh38 | Gain |
|
| esv3568885 | Chr.3:87182571 - 87518398 on Build GRCh38 | Loss |
|
| nsv470749 | Chr.3:87258397 - 87302230 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map