Assay Details
Target Gene Details
Entrez Gene ID: | 100505877 |
Gene Name: | UBE2E2 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.3:23195070-23202993 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE2E2-AS1 | NR_136176.1 | |||
| NR_136177.1 | ||||
| AI990151.1 | ||||
| HY029877.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834636 | Chr.3:23097162 - 23259952 on Build GRCh38 | Gain+Loss |
|
| nsv1005186 | Chr.3:21989661 - 24089392 on Build GRCh38 | Gain |
|
| nsv3731 | Chr.3:23173270 - 23207099 on Build GRCh38 | Insertion |
|

Genomic Map