Assay Details
Target Gene Details
Entrez Gene ID: | 339896 |
Gene Name: | glutamate decarboxylase like 1 |
Gene Aliases: |
ADC, CSADC, HuADC, HuCSADC |
Location: |
Chr.3:30726009-30894661 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GADL1 | NM_207359.2 | NP_997242.2 | ||
| XM_017006297.1 | XP_016861786.1 | |||
| XM_017006298.1 | XP_016861787.1 | |||
| AK128643.1 | BAC87546.1 | |||
| BC093701.1 | AAH93701.1 | |||
| BC111986.1 | AAI11987.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv590029 | Chr.3:30744939 - 30761548 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map