Assay Details
Target Gene Details
Entrez Gene ID: | 25819 |
Gene Name: | nocturnin |
Gene Aliases: |
CCR4L, CCRN4L, Ccr4c, NOC |
Location: |
Chr.4:139015759-139045939 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOCT | NM_012118.3 | NP_036250.2 | ||
| AF183961.1 | AAD56548.1 | |||
| BC021963.2 | AAH21963.1 | |||
| BC023512.2 | AAH23512.1 | |||
| BC077723.1 | AAH77723.1 | |||
| BC113494.1 | ||||
| BC113500.1 | ||||
| HY067201.1 |
More Information
Set Membership: |
|

Genomic Map