Assay Details
Target Gene Details
Entrez Gene ID: | 93587 |
Gene Name: | tRNA methyltransferase 10A |
Gene Aliases: |
HEL-S-88, MSSGM, MSSGM1, RG9MTD2, TRM10 |
Location: |
Chr.4:99546707-99564057 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRMT10A | NM_001134665.2 | NP_001128137.1 | ||
| NM_001134666.2 | NP_001128138.1 | |||
| NM_152292.4 | NP_689505.1 | |||
| XM_005263352.3 | XP_005263409.1 | |||
| XM_006714417.2 | XP_006714480.1 | |||
| AF106046.1 | AAD21019.1 | |||
| AK000332.1 | ||||
| AK313548.1 | ||||
| BC028373.2 | AAH28373.1 | |||
| EU668342.1 |
More Information
Set Membership: |
|

Genomic Map