Assay Details
Target Gene Details
Entrez Gene ID: | 10886 |
Gene Name: | neuropeptide FF receptor 2 |
Gene Aliases: |
GPR74, HLWAR77, NPFF2, NPGPR |
Location: |
Chr.4:72031804-72148201 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPFFR2 | NM_004885.2 | NP_004876.2 | ||
| AF119815.1 | AAD22047.1 | |||
| AJ311393.1 | CAC85427.1 | |||
| AK308069.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3569516 | Chr.4:72018593 - 72038075 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78760772] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map