Assay Details
Target Gene Details
Entrez Gene ID: | 9407 |
Gene Name: | transmembrane protease, serine 11D |
Gene Aliases: |
HAT |
Location: |
Chr.4:67820876-67885063 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMPRSS11D | NM_004262.2 | NP_004253.1 | ||
| XM_005265710.4 | XP_005265767.1 | |||
| XM_017008851.1 | XP_016864340.1 | |||
| AB002134.1 | BAA28691.1 | |||
| AK094054.1 | ||||
| AK290289.1 | ||||
| AK296853.1 | ||||
| BC125195.1 | ||||
| BC125196.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829960 | Chr.4:67783519 - 67943911 on Build GRCh38 | Loss |
|
| esv3600854 | Chr.4:67821801 - 67827581 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map