Assay Details
Target Gene Details
Entrez Gene ID: | 79071 |
Gene Name: | ELOVL fatty acid elongase 6 |
Gene Aliases: |
FACE, FAE, LCE |
Location: |
Chr.4:110049073-110199199 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELOVL6 | NM_001130721.1 | NP_001124193.1 | ||
| NM_024090.2 | NP_076995.1 | |||
| XM_011532233.2 | XP_011530535.1 | |||
| XM_011532234.2 | XP_011530536.1 | |||
| XM_011532235.2 | XP_011530537.1 | |||
| AK027031.1 | BAB15632.1 | |||
| AK074813.1 | BAC11225.1 | |||
| BC001305.1 | AAH01305.1 | |||
| BP314858.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv595145 | Chr.4:110048416 - 110061092 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map