Assay Details
Target Gene Details
Entrez Gene ID: | 55858 |
Gene Name: | transmembrane protein 165 |
Gene Aliases: |
CDG2K, FT27, GDT1, TMPT27, TPARL |
Location: |
Chr.4:55395913-55453397 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM165 | NM_018475.4 | NP_060945.2 | ||
| NR_073070.1 | ||||
| XM_011534394.2 | XP_011532696.1 | |||
| XM_017008412.1 | XP_016863901.1 | |||
| AF132746.1 | AAL75947.1 | |||
| AF183409.1 | AAG09678.1 | |||
| AF220188.1 | AAF67653.1 | |||
| AK290663.1 | ||||
| AK295289.1 | ||||
| BC104978.1 | AAI04979.1 | |||
| BC104980.1 | AAI04981.1 | |||
| BC107582.1 | AAI07583.1 | |||
| DA371259.1 | ||||
| HY089602.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2660719 | Chr.4:55399828 - 55403725 on Build GRCh38 | Deletion |
|
| nsv1131801 | Chr.4:55403079 - 55403335 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map