Assay Details
Target Gene Details
Entrez Gene ID: | 4258 |
Gene Name: | microsomal glutathione S-transferase 2 |
Gene Aliases: |
GST2, MGST-II |
Location: |
Chr.4:139665725-139754546 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MGST2 | NM_001204366.1 | NP_001191295.1 | ||
| NM_001204367.1 | NP_001191296.1 | |||
| NM_001204368.1 | NP_001191297.1 | |||
| NM_002413.4 | NP_002404.1 | |||
| XM_006714221.2 | XP_006714284.1 | |||
| XM_017008211.1 | XP_016863700.1 | |||
| XM_017008212.1 | XP_016863701.1 | |||
| XM_017008213.1 | XP_016863702.1 | |||
| XM_017008214.1 | XP_016863703.1 | |||
| AA122237.1 | ||||
| AK130948.1 | ||||
| AK307231.1 | ||||
| BC025416.1 | AAH25416.1 | |||
| BE312976.1 | ||||
| CR407640.1 | CAG28568.1 | |||
| DB257575.1 | ||||
| U77604.1 | AAC51768.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs114596735] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map