Assay Details
Target Gene Details
Entrez Gene ID: | 105377348 |
Gene Name: | uncharacterized LOC105377348 |
Gene Aliases: |
- |
Location: |
Chr.4:102828055-102844106 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105377348 | NR_131185.1 | |||
| NR_131186.1 | ||||
| AK093356.1 | ||||
| CD368268.1 | ||||
| HY045516.1 |
Target Gene Details
Entrez Gene ID: | 7323 |
Gene Name: | ubiquitin conjugating enzyme E2 D3 |
Gene Aliases: |
E2(17)KB3, UBC4/5, UBCH5C |
Location: |
Chr.4:102794383-102868893 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE2D3 | NM_181890.2 | NP_871619.1 | ||
| NM_181893.2 | NP_871622.1 | |||
| BG613555.1 | ||||
| BU570661.1 | ||||
| DA965437.1 | ||||
| HY109217.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv594957 | Chr.4:102806853 - 102948934 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78625384] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map