Assay Details
Target Gene Details
Entrez Gene ID: | 401136 |
Gene Name: | TMPRSS11B N-terminal like, pseudogene |
Gene Aliases: |
- |
Location: |
Chr.4:68184129-68218080 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMPRSS11BNL | NR_104048.1 | |||
| AK123556.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv594506 | Chr.4:68183925 - 68357339 on Build GRCh38 | Loss |
|

Genomic Map