Assay Details
Target Gene Details
Entrez Gene ID: | 2044 |
Gene Name: | EPH receptor A5 |
Gene Aliases: |
CEK7, EHK-1, EHK1, EK7, HEK7, TYRO4 |
Location: |
Chr.4:65319563-65670838 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPHA5 | NM_001281765.2 | 18 | 7366 | NP_001268694.1 |
| NM_001281766.2 | 17 | 7300 | NP_001268695.1 | |
| NM_001318761.1 | 18 | 6484 | NP_001305690.1 | |
| NM_004439.7 | 18 | 7363 | NP_004430.4 | |
| NM_182472.4 | 17 | 7297 | NP_872272.2 | |
| XM_005265653.3 | 16 | 7217 | XP_005265710.1 | |
| XM_011531735.2 | 17 | 7373 | XP_011530037.1 | |
| XM_017007878.1 | 16 | 7307 | XP_016863367.1 | |
| XM_017007880.1 | 15 | 7151 | XP_016863369.1 | |
| BX537946.1 | 18 | 6485 | CAD97914.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470041 | Chr.4:65311056 - 65392555 on Build GRCh38 | Loss |
|
| nsv1014463 | Chr.4:65294286 - 65989398 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map