Assay Details
Target Gene Details
Entrez Gene ID: | 84708 |
Gene Name: | ligand of numb-protein X 1 |
Gene Aliases: |
LNX, MPDZ, PDZRN2 |
Location: |
Chr.4:53458264-53652524 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LNX1 | NM_001126328.2 | NP_001119800.1 | ||
| NM_032622.2 | NP_116011.2 | |||
| XM_005265785.4 | XP_005265842.1 | |||
| XM_017008776.1 | XP_016864265.1 | |||
| AF237782.1 | AAK15039.1 | |||
| AK056823.1 | BAB71291.1 | |||
| AK294611.1 | ||||
| AY358326.1 | AAQ88692.1 | |||
| BC022983.1 | AAH22983.1 | |||
| BC034737.1 | AAH34737.1 |
Target Gene Details
Entrez Gene ID: | 100873939 |
Gene Name: | LNX1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.4:53500077-53523274 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LNX1-AS1 | NR_046622.1 | |||
| AA405946.1 | ||||
| BX103714.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829936 | Chr.4:53501616 - 53689956 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map