Assay Details
Target Gene Details
Entrez Gene ID: | 2920 |
Gene Name: | C-X-C motif chemokine ligand 2 |
Gene Aliases: |
CINC-2a, GRO2, GROb, MGSA-b, MIP-2a, MIP2, MIP2A, SCYB2 |
Location: |
Chr.4:74097035-74099280 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CXCL2 | NM_002089.3 | NP_002080.1 | ||
| AF043340.1 | AAC03540.1 | |||
| BC015753.1 | AAH15753.1 | |||
| BX334969.2 | ||||
| CR542171.1 | CAG46968.1 | |||
| M36820.1 | AAA63183.1 | |||
| M57731.1 | AAA63182.1 | |||
| X53799.1 | CAA37808.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1006029 | Chr.4:73928205 - 74120947 on Build GRCh38 | Gain |
|
| esv3575747 | Chr.4:73914851 - 74124725 on Build GRCh38 | Gain |
|
| nsv525980 | Chr.4:73865809 - 74556070 on Build GRCh38 | Gain |
|
| nsv1004768 | Chr.4:73908655 - 74243862 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map