Assay Details
Target Gene Details
Entrez Gene ID: | 2560 |
Gene Name: | gamma-aminobutyric acid type A receptor beta1 subunit |
Gene Aliases: |
- |
Location: |
Chr.4:47031278-47438409 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GABRB1 | NM_000812.3 | NP_000803.2 | ||
| XM_017007986.1 | XP_016863475.1 | |||
| AK225167.1 | ||||
| AK225686.1 | ||||
| AK293691.1 | ||||
| AK312720.1 | ||||
| BC022449.1 | AAH22449.1 | |||
| KJ534841.1 | ||||
| KJ535054.1 | ||||
| X14767.1 | CAA32875.1 |
More Information
Set Membership: |
|

Genomic Map