Assay Details
Target Gene Details
Entrez Gene ID: | 3033 |
Gene Name: | hydroxyacyl-CoA dehydrogenase |
Gene Aliases: |
HAD, HADH1, HADHSC, HCDH, HHF4, MSCHAD, SCHAD |
Location: |
Chr.4:107989714-108035175 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HADH | NM_001184705.2 | NP_001171634.2 | ||
NM_005327.4 | NP_005318.3 | |||
XM_005262972.2 | XP_005263029.1 | |||
AF001902.1 | AAB54008.1 | |||
AF001903.1 | AAB54009.1 | |||
AK096018.1 | ||||
AK314444.1 | ||||
BC000306.2 | AAH00306.1 | |||
DA825380.1 | ||||
X96752.1 | CAA65528.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1003893 | Chr.4:107993422 - 108023690 on Build GRCh38 | Gain |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
