Assay Details
Target Gene Details
Entrez Gene ID: | 56477 |
Gene Name: | C-C motif chemokine ligand 28 |
Gene Aliases: |
CCK1, MEC, SCYA28 |
Location: |
Chr.5:43356972-43412391 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCL28 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508352 | Chr.5:43237728 - 43390960 on Build GRCh38 | Deletion |
|
| esv3604897 | Chr.5:43343819 - 43405322 on Build GRCh38 | Gain |
|

Genomic Map