Assay Details
Target Gene Details
Entrez Gene ID: | 28966 |
Gene Name: | sorting nexin 24 |
Gene Aliases: |
PRO1284, SBBI31 |
Location: |
Chr.5:122845465-123029318 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNX24 | NM_014035.2 | NP_054754.1 | ||
| XM_005271972.4 | XP_005272029.1 | |||
| XM_006714592.3 | XP_006714655.1 | |||
| XM_011543349.2 | XP_011541651.1 | |||
| XM_011543350.2 | XP_011541652.1 | |||
| XM_011543352.1 | XP_011541654.1 | |||
| XM_017009395.1 | XP_016864884.1 | |||
| XM_017009396.1 | XP_016864885.1 | |||
| AF139461.1 | AAD32668.1 | |||
| AY044655.1 | AAK98769.1 | |||
| AY358098.1 | AAQ88465.1 | |||
| BC010886.1 | AAH10886.1 | |||
| BC069012.1 | AAH69012.1 | |||
| BQ278046.1 | ||||
| DB498436.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830471 | Chr.5:122796424 - 122988304 on Build GRCh38 | Gain |
|
| esv3606594 | Chr.5:122852505 - 122855233 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map