Assay Details
Target Gene Details
Entrez Gene ID: | 101928794 |
Gene Name: | uncharacterized LOC101928794 |
Gene Aliases: |
- |
Location: |
Chr.5:67001618-67004154 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928794 | NR_111908.1 | 1 | 204 | |
| AW003767.1 | 1 | 204 | ||
| AW138729.1 | 1 | 262 | ||
| BQ011784.1 |
Target Gene Details
Entrez Gene ID: | 375449 |
Gene Name: | microtubule associated serine/threonine kinase family member 4 |
Gene Aliases: |
- |
Location: |
Chr.5:66596348-67169595 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAST4 | NM_001164664.1 | NP_001158136.1 | ||
| NM_001290226.1 | NP_001277155.1 | |||
| NM_015183.2 | NP_055998.1 | |||
| XM_011543382.2 | XP_011541684.1 | |||
| XM_011543385.2 | XP_011541687.1 | |||
| XM_017009447.1 | XP_016864936.1 | |||
| XM_017009448.1 | XP_016864937.1 | |||
| XM_017009449.1 | XP_016864938.1 | |||
| XM_017009450.1 | XP_016864939.1 | |||
| XM_017009451.1 | XP_016864940.1 | |||
| XM_017009452.1 | XP_016864941.1 | |||
| AK131421.1 | ||||
| AK299538.1 | ||||
| AY830839.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1023631 | Chr.5:66918448 - 67054126 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map