Assay Details
Target Gene Details
Entrez Gene ID: | 6793 |
Gene Name: | serine/threonine kinase 10 |
Gene Aliases: |
LOK, PRO2729 |
Location: |
Chr.5:172042070-172188342 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STK10 | NM_005990.3 | 19 | 4634 | NP_005981.3 |
| XM_017009788.1 | 18 | 4188 | XP_016865277.1 | |
| AK092814.1 | 2 | 1840 |
More Information
Set Membership: |
|

Genomic Map