Assay Details
Target Gene Details
Entrez Gene ID: | 153328 |
Gene Name: | solute carrier family 25 member 48 |
Gene Aliases: |
- |
Location: |
Chr.5:135579002-135888637 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A48 | XM_017009090.1 | XP_016864579.1 | ||
| XM_017009092.1 | XP_016864581.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs79030846] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map