Assay Details
Target Gene Details
Entrez Gene ID: | 2533 |
Gene Name: | FYN binding protein |
Gene Aliases: |
ADAP, PRO0823, SLAP-130, SLAP130 |
Location: |
Chr.5:39105252-39274523 on Build GRCh38 |
Assay Gene Location: | Within Intron 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FYB | NM_001243093.1 | NP_001230022.1 | ||
| NM_001465.4 | NP_001456.3 | |||
| NM_199335.3 | NP_955367.1 | |||
| XM_006714464.2 | XP_006714527.1 | |||
| XM_006714465.2 | XP_006714528.1 | |||
| XM_006714466.2 | XP_006714529.1 | |||
| XM_011514008.2 | XP_011512310.1 | |||
| XM_011514009.1 | XP_011512311.1 | |||
| XM_011514010.1 | XP_011512312.1 | |||
| XM_011514011.2 | XP_011512313.1 | |||
| XM_017009316.1 | XP_016864805.1 | |||
| AF001862.1 | AAB62226.1 | |||
| AF198052.1 | AAF62400.1 | |||
| AK290403.1 | ||||
| AK297077.1 | ||||
| BC117449.1 | ||||
| BC143645.1 | ||||
| BX647195.1 | ||||
| BX647196.1 | ||||
| CD701507.1 | ||||
| U93049.1 | AAC51300.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4804 | Chr.5:39090985 - 39125155 on Build GRCh38 | Insertion |
|
| nsv529023 | Chr.5:38955694 - 39119621 on Build GRCh38 | Loss |
|
| nsv950671 | Chr.5:39022399 - 39283598 on Build GRCh38 | Duplication |
|
| nsv597860 | Chr.5:38995581 - 40274356 on Build GRCh38 | Gain |
|
| nsv4805 | Chr.5:39095098 - 39122592 on Build GRCh38 | Deletion |
|
| nsv597871 | Chr.5:39105492 - 39123914 on Build GRCh38 | Loss |
|
| esv3604776 | Chr.5:39072365 - 39193316 on Build GRCh38 | Loss |
|
| esv1006514 | Chr.5:39108035 - 39111190 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map