Assay Details
Target Gene Details
Entrez Gene ID: | 7265 |
Gene Name: | tetratricopeptide repeat domain 1 |
Gene Aliases: |
TPR1 |
Location: |
Chr.5:160009100-160065545 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC1 | NM_001282500.1 | NP_001269429.1 | ||
| NM_003314.2 | NP_003305.1 | |||
| AK222641.1 | BAD96361.1 | |||
| AK222984.1 | BAD96704.1 | |||
| AK315262.1 | ||||
| BP245572.1 | ||||
| DN989835.1 | ||||
| U46570.1 | AAB36871.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830531 | Chr.5:159896337 - 160076045 on Build GRCh38 | Loss |
|
| nsv1015634 | Chr.5:159750943 - 160620377 on Build GRCh38 | Gain |
|
| dgv1150e214 | Chr.5:159917404 - 160051796 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map