Assay Details
Target Gene Details
Entrez Gene ID: | 7025 |
Gene Name: | nuclear receptor subfamily 2 group F member 1 |
Gene Aliases: |
BBOAS, BBSOAS, COUP-TFI, EAR-3, EAR3, ERBAL3, NR2F2, SVP44, TCFCOUP1, TFCOUP1 |
Location: |
Chr.5:93583337-93594613 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NR2F1 | NM_005654.5 | NP_005645.1 | ||
| BC004154.2 | AAH04154.1 | |||
| BC017493.1 | AAH17493.1 | |||
| HQ709180.1 | ||||
| HQ709181.1 | ||||
| HQ709182.1 | ||||
| HQ709183.1 | ||||
| X16155.1 | CAA34277.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4918 | Chr.5:93562720 - 93607653 on Build GRCh38 | Deletion |
|
| nsv598949 | Chr.5:93584409 - 93587075 on Build GRCh38 | Loss |
|
| dgv954n67 | Chr.5:93560640 - 93633962 on Build GRCh38 | Gain |
|
| nsv830404 | Chr.5:93532630 - 93712551 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map