Assay Details
Target Gene Details
Entrez Gene ID: | 55255 |
Gene Name: | WD repeat domain 41 |
Gene Aliases: |
MSTP048 |
Location: |
Chr.5:77426652-77620611 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WDR41 | NM_018268.3 | NP_060738.2 | ||
| XM_005248551.4 | XP_005248608.1 | |||
| XM_011543505.2 | XP_011541807.1 | |||
| XM_017009606.1 | XP_016865095.1 | |||
| AF115511.1 | AAO06948.1 | |||
| AK001766.1 | BAA91895.1 | |||
| AK021855.1 | BAB13915.1 | |||
| AK023977.1 | BAB14749.1 | |||
| AK293316.1 | ||||
| AK300058.1 | ||||
| AK304329.1 | ||||
| AL834138.1 | CAD38853.2 | |||
| BC040241.1 | AAH40241.1 | |||
| KU178690.1 | ||||
| KU178691.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs78666564] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map