Assay Details
Target Gene Details
Entrez Gene ID: | 9879 |
Gene Name: | DEAD-box helicase 46 |
Gene Aliases: |
PRPF5, Prp5 |
Location: |
Chr.5:134758734-134831138 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX46 | NM_001300860.1 | NP_001287789.1 | ||
| NM_014829.3 | NP_055644.2 | |||
| NR_125341.1 | ||||
| XM_011543758.1 | XP_011542060.1 | |||
| AB018344.1 | ||||
| AF106680.1 | AAD43033.1 | |||
| AK126011.1 | ||||
| AK291788.1 | ||||
| BC012304.2 | AAH12304.1 | |||
| BC105598.1 | ||||
| BF244218.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575882 | Chr.5:134672959 - 134779811 on Build GRCh38 | Gain |
|
| nsv830493 | Chr.5:134667777 - 134926221 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map