Assay Details
Target Gene Details
Entrez Gene ID: | 91368 |
Gene Name: | CDKN2A interacting protein N-terminal like |
Gene Aliases: |
- |
Location: |
Chr.5:134402065-134414625 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDKN2AIPNL | NM_080656.2 | NP_542387.1 | ||
| BC008293.1 | AAH08293.1 |
More Information
Set Membership: |
|

Genomic Map