Assay Details
Target Gene Details
Entrez Gene ID: | 133396 |
Gene Name: | interleukin 31 receptor A |
Gene Aliases: |
CRL, CRL3, GLM-R, GLMR, GPL, IL-31RA, PLCA2, PRO21384, hGLM-R |
Location: |
Chr.5:55840334-55922854 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL31RA | NM_001242637.1 | NP_001229566.1 | ||
| NM_001242638.1 | NP_001229567.1 | |||
| NM_001297570.1 | NP_001284499.1 | |||
| NM_001297572.1 | NP_001284501.1 | |||
| NM_139017.5 | NP_620586.3 | |||
| XM_011543142.2 | XP_011541444.1 | |||
| AK297938.1 | ||||
| AY358740.1 | AAQ89100.1 | |||
| AY499339.1 | AAS86444.1 | |||
| AY499340.1 | AAS86445.1 | |||
| AY499341.1 | AAS86446.1 | |||
| AY499342.1 | AAS86447.1 |
More Information
Set Membership: |
|

Genomic Map