Assay Details
Target Gene Details
Entrez Gene ID: | 100861519 |
Gene Name: | GDNF antisense RNA 1 (head to head) |
Gene Aliases: |
GDNFOS |
Location: |
Chr.5:37840028-37875799 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GDNF-AS1 | NM_001278094.1 | 1 | 545 | NP_001265023.1 |
| NR_103441.1 | ||||
| NR_103442.1 | ||||
| XM_017008916.1 | XP_016864405.1 | |||
| XM_017008917.1 | XP_016864406.1 | |||
| XM_017008918.1 | XP_016864407.1 | |||
| DA359107.1 | 1 | 251 | ||
| JF824129.1 | 1 | 545 | ||
| JF824130.1 | ||||
| JF824131.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3604753 | Chr.5:37854044 - 37997903 on Build GRCh38 | Gain |
|

Genomic Map