Assay Details
Target Gene Details
Entrez Gene ID: | 1462 |
Gene Name: | versican |
Gene Aliases: |
CSPG2, ERVR, GHAP, PG-M, WGN, WGN1 |
Location: |
Chr.5:83471674-83582303 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VCAN | NM_001126336.2 | NP_001119808.1 | ||
| NM_001164097.1 | NP_001157569.1 | |||
| NM_001164098.1 | NP_001157570.1 | |||
| NM_004385.4 | NP_004376.2 | |||
| AB209491.1 | BAD92728.1 | |||
| AK307642.1 | ||||
| BC050524.1 | AAH50524.1 | |||
| BP332680.1 | ||||
| D32039.1 | BAA06801.1 | |||
| DA361670.1 | ||||
| U16306.1 | AAA65018.1 | |||
| U26555.1 | AAA67565.1 | |||
| X15998.1 | CAA34128.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830366 | Chr.5:83335870 - 83517681 on Build GRCh38 | Gain |
|
| nsv598784 | Chr.5:83442024 - 83480087 on Build GRCh38 | Loss |
|
| nsv1015852 | Chr.5:83404850 - 84499122 on Build GRCh38 | Gain |
|
| nsv4903 | Chr.5:83440906 - 83474949 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map