Assay Details
Target Gene Details
Entrez Gene ID: | 1452 |
Gene Name: | casein kinase 1 alpha 1 |
Gene Aliases: |
CK1, CK1a, CKIa, HEL-S-77p, HLCDGP1, PRO2975 |
Location: |
Chr.5:149495894-149551552 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CSNK1A1 | NM_001025105.2 | NP_001020276.1 | ||
| NM_001271741.1 | NP_001258670.1 | |||
| NM_001271742.1 | NP_001258671.1 | |||
| NM_001892.5 | NP_001883.4 | |||
| AF218004.1 | AAG17246.1 | |||
| AF447582.1 | AAL60204.1 | |||
| AK293761.1 | ||||
| AK294099.1 | ||||
| AK294942.1 | ||||
| AK303794.1 | ||||
| BC008717.2 | AAH08717.1 | |||
| BX647478.1 | ||||
| BX648552.1 | ||||
| L37042.1 | AAC41760.1 | |||
| X80693.1 | CAA56710.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv24490 | Chr.5:149440562 - 149595691 on Build GRCh38 | Gain |
|
| nsv516848 | Chr.5:149490537 - 149522775 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map