Assay Details
Target Gene Details
Entrez Gene ID: | 10087 |
Gene Name: | collagen type IV alpha 3 binding protein |
Gene Aliases: |
CERT, CERTL, GPBP, MRD34, STARD11 |
Location: |
Chr.5:75368486-75511981 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COL4A3BP | NM_001130105.1 | NP_001123577.1 | ||
| NM_005713.2 | NP_005704.1 | |||
| NM_031361.2 | NP_112729.1 | |||
| XM_006714513.2 | XP_006714576.1 | |||
| XM_011543090.2 | XP_011541392.1 | |||
| XM_011543091.2 | XP_011541393.1 | |||
| XM_017008919.1 | XP_016864408.1 | |||
| AK000604.1 | ||||
| AK091851.1 | BAC03762.1 | |||
| AK096854.1 | ||||
| AK292087.1 | ||||
| BC000102.2 | AAH00102.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575856 | Chr.5:75260838 - 75400813 on Build GRCh38 | Gain |
|
| nsv980685 | Chr.5:75374056 - 75374913 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map