Assay Details
Target Gene Details
Entrez Gene ID: | 3567 |
Gene Name: | interleukin 5 |
Gene Aliases: |
EDF, IL-5, TRF |
Location: |
Chr.5:132539194-132556827 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL5 | XM_005271988.3 | 5 | 1542 | XP_005272045.1 |
| XM_011543373.2 | 6 | 1644 | XP_011541675.1 | |
| XM_011543374.2 | 5 | 1595 | XP_011541676.1 |
More Information
Set Membership: |
|

Genomic Map