Assay Details
Target Gene Details
Entrez Gene ID: | 55255 |
Gene Name: | WD repeat domain 41 |
Gene Aliases: |
MSTP048 |
Location: |
Chr.5:77426652-77620611 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WDR41 | NM_018268.3 | 13 | 3098 | NP_060738.2 |
| XM_005248551.4 | 11 | 2891 | XP_005248608.1 | |
| XM_011543505.2 | 13 | 2860 | XP_011541807.1 | |
| XM_017009606.1 | 13 | 3508 | XP_016865095.1 | |
| BG113002.1 | ||||
| CD656920.1 | 1 | 373 |

Genomic Map