Assay Details
Target Gene Details
Entrez Gene ID: | 2150 |
Gene Name: | F2R like trypsin receptor 1 |
Gene Aliases: |
GPR11, PAR2 |
Location: |
Chr.5:76819008-76835315 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
F2RL1 | XM_017009223.1 | XP_016864712.1 | ||
AY336105.1 | AAP97012.1 | |||
BC012453.1 | AAH12453.1 | |||
BC018130.1 | AAH18130.1 | |||
BT009856.1 | AAP88858.1 | |||
DB463838.1 | ||||
U34038.1 | AAB47871.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv5703n100 | Chr.5:76783895 - 76838952 on Build GRCh38 | Gain |
![]() |
esv3605474 | Chr.5:76789709 - 76840353 on Build GRCh38 | Gain |
![]() |
esv2763908 | Chr.5:76787569 - 76838952 on Build GRCh38 | Gain |
![]() |
dgv9885n54 | Chr.5:76790772 - 76834288 on Build GRCh38 | Gain |
![]() |
esv3575858 | Chr.5:76790970 - 76832928 on Build GRCh38 | Gain |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
